Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EPHA2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7554R-FITC
BOSSBS-7554R-FITCEA 930 EUR InStock
BOSSBS-7554R-FITC
Anti-EPHA2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Eph receptor A2 is the receptor for members of the ephrin-A family. It binds to ephrin-A1, -A3, -A4 and -A5. Eph receptor A3 is also a receptor for members of the ephrin-A family. It binds to ephrin-A2, -A3, -A4 and -A5 and is thought to play a role in lymphoid function. Eph receptor A4 is a receptor for members of the ephrin-A family. It binds to ephrin-A1, -A4 and -A5. It binds more poorly to ephrin-A2 and -A3. It may play a role in a signal transduction process involved in hindbrain pattern formation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EPHA2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge