Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TNIP2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7563R-CY5
BOSSBS-7563R-CY5EA 900 EUR InStock
BOSSBS-7563R-CY5
Anti-TNIP2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Inhibits NF-kappa-B activation by blocking the interaction of RIPK1 with its downstream effector NEMO/IKBKG. Forms a ternary complex with NFKB1 and MAP3K8 but appears to function upstream of MAP3K8 in the TLR4 signaling pathway that regulates MAP3K8 activation. Involved in activation of the MEK/ERK signaling pathway during innate immune response; this function seems to be stimulus- and cell type specific. Required for stability of MAP3K8. Involved in regulation of apoptosis in endothelial cells; promotes TEK agonist-stimulated endothelial survival. May act as transcriptional coactivator when translocated to the nucleus. Enhances CHUK-mediated NF-kappa-B activation involving NF-kappa-B p50-p65 and p50-c-Rel complexes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNIP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge