Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ASB3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7736R-CY5.5
BOSSBS-7736R-CY5.5EA 900 EUR InStock
BOSSBS-7736R-CY5.5
Anti-ASB3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Members of the suppressor of cytokine signaling (SOCS) family of proteins contain C-terminal regions of homology called the SOCS box, which serves to couple SOCS proteins and their binding partners with the elongin B and C complex. Serveral other families of proteins also contain SOCS boxes but differ from the SOCS proteins in the type of domain they contained upstream of the SOCS box. Four members of the ankyrin repeat and SOCS box-containing (ASB) protein family are identified and termed as ASB-1, ASB-2, ASB-3 and ASB-4. ASB-1 is expressed in multiple organs, including the hematopoietic compartment. ASB-1 knock-out mice display a diminution of spermatogenesis with less complete filling of seminiferous tubules. Asb-2 is a novel retinoic-acid (RA)-induced gene in acute promyelocytic leukemia (APL) cells and its expression induces growth-inhibition and chromatin condensation recapitulating early events critical to RA-induced differentiaiton of APL cells. ASB-2 is directly induced by all-trans retinotic acid, by the binding of RARa to the RAR binding element/RXR binding element in the Asb-2 promoter.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ASB3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge