Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KLHL13 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7752R-A350
BOSSBS-7752R-A350EA 893 EUR InStock
BOSSBS-7752R-A350
Anti-KLHL13 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
KLHL13 (kelch like 13), also known as BKLHD2 (BTB and kelch domain containing protein 2), is a 604 amino acid protein that contains six Kelch repeats and one BTB/POZ domain. Expressed predominantly in brain, KLHL13 is believed to play a role in protein ubiquitination and may function as a substrate specific adapter of an E3 ubiquitin protein ligase complex. E3 ligases accept a ubiquitin residue from an E2 ubiquitin conjugating enzyme and immediately transfer that residue to a protein that is targeted for degradation. Specifically, KLHL13 interacts with KLHL9 and CUL3, a member of the cullin family of mediators that participate in the selective targeting of proteins for ubiquitin mediated proteolysis. Via its BTB and C terminal Kelch (BACK) motif, KLHL13 is thought to play a role in spatially orientating substrates in the CUL3 ligase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLHL13
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge