Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DSN1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7796R-A488
BOSSBS-7796R-A488EA 1350 EUR InStock
BOSSBS-7796R-A488
Anti-DSN1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
This gene encodes a kinetochore protein that functions aspart of the minichromosome instability-12 centromere complex. Theencoded protein is required for proper kinetochore assembly andprogression through the cell cycle. Alternative splicing results inmultiple transcript variants. [provided by RefSeq, Feb 2009].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DSN1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge