Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SKA2 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7847R
BOSSBS-7847REA 725 EUR InStock
BOSSBS-7847R
Anti-SKA2 Rabbit Polyclonal Antibody
Antikörper
Ska2 (spindle and kinetochore associated complex subunit 2), also known as FAM33A, is a 121 amino acid component of the Ska1 complex, a microtubule-binding subcomplex of the outer kinetochore that is critical for proper chromosome segregation. The Ska1 complex is a component of the kinetochore-microtubule interface and directly associates with microtubules as oligomeric assemblies. Localized to the outer kinetochore and spindle microtubules during cell proliferation, Ska2 is essential for spindle checkpoint silencing and exit from mitosis. Downregulation of Ska2 leads to delayed recruitment of MAD2, a component of the mitotic spindle checkpoint, to several kinetochores resulting in occasional loss of individual chromosomes from the metaphase plate. Ska2 is encoded by a gene located on human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SKA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge