Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KATNB1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7851R-CY5.5
BOSSBS-7851R-CY5.5EA 900 EUR InStock
BOSSBS-7851R-CY5.5
Anti-KATNB1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Microtubules, polymers of alpha and beta tubulin subunits,form the mitotic spindle of a dividing cell and help to organizemembranous organelles during interphase. Katanin is a heterodimerthat consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDaaccessory protein (p80 subunit B 1). The p60 subunit acts to severand disassemble microtubules, while the p80 subunit targets theenzyme to the centrosome. Katanin is a member of the AAA family ofATPases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KATNB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge