Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ALOXE3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7856R-HRP
BOSSBS-7856R-HRPEA 900 EUR InStock
BOSSBS-7856R-HRP
Anti-ALOXE3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Introduces molecular oxygen into polyunsaturated fatty acids. Exact substrate is not known.Tissue specificity:Predominantly expressed in skin.Involvement in diseaseDefects in ALOXE3 are a cause of non-bullous congenital ichthyosiform erythroderma (NCIE). NCIE is a non-bullous ichthyosis, a skin disorder characterized by abnormal cornification of the epidermis. Most affected individuals are born with a tight, shiny, translucent covering called collodion membrane. The collodion membrane subsequently evolves into generalized scaling and intense redness of the skin. Clinical features are milder than in lamellar ichthyoses and demonstrate a greater variability in the intensity of erythema, size and type of scales. In contrast to lamellar ichthyoses, scales are usually white, fine and powdery, and palms and soles are severely affected. Patients suffer from palmoplantar keratoderma, often with painful fissures, digital contractures, and loss of pulp volume.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ALOXE3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge