Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FAM3C Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7862R-CY7
BOSSBS-7862R-CY7EA 900 EUR InStock
BOSSBS-7862R-CY7
Anti-FAM3C Rabbit Polyclonal Antibody (Cy7®)
Antikörper
ILEI is a 227 amino acid, ubiquitously expressed protein containing an amino-terminal signal peptide. Elevated levels of ILEI translation are observed in oncogenic, Ras-transformed mammary epithelial cells and epithelial to mesenchymal transition (Emt) as well as tumor growth and metastasis. Also, overexpression of ILEI results in loss of ZO-1, a protein involved in tight junctions, and expression of cytoplasmic E-cadherin, which has been shown to influence loss of polarity and invasiveness. Due to this evidence, it is suspected that ILEI cooperates with oncogenic Ras to cause TGF∫-independent Emt and its overexpression is correlated with the invasion, metastasis and survival in a variety of cancers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM3C
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge