Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ROPN1L Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7903R-A647
BOSSBS-7903R-A647EA 893 EUR InStock
BOSSBS-7903R-A647
Anti-ROPN1L Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
AKAP associated sperm protein (encoded by the gene ROPN1L) is a sperm protein which interacts with A-kinase anchoring protein, AKAP3, through the amphipathic helix region of AKAP3. The Type II regulatory subunit of cAMP-dependent protein kinase (PKARII) also binds to this helical domain of AKAP3, allowing PKARII to be targeted to specific subcellular compartments. It is suggested that sperm contains several proteins that bind to AKAPs in a manner similar to PKARII, and this encoded protein may be one of them.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ROPN1L
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge