Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CA7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7984R-A750
BOSSBS-7984R-A750EA 893 EUR InStock
BOSSBS-7984R-A750
Anti-CA7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Carbonic anhydrases are a large family of zincmetalloenzymes that catalyze the reversible hydration of carbondioxide. They participate in a variety of biological processes,including respiration, calcification, acid-base balance, boneresorption, and the formation of aqueous humor, cerebrospinalfluid, saliva, and gastric acid. They show extensive diversity intissue distribution and in their subcellular localisation. Thecytosolic protein encoded by this gene is predominantly expressedin the salivary glands. Alternative splicing in the coding regionresults in multiple transcript variants encoding differentisoforms.

Type: Primary
Antigen: CA7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 86-150/264
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge