Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-COLEC11 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8032R-CY7
BOSSBS-8032R-CY7EA 900 EUR InStock
BOSSBS-8032R-CY7
Anti-COLEC11 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
COLEC11 is a 271 amino acid C-type lectin protein that contains a collagen-like domain and a carbohydrate recognition domain, and plays an important role in host-defense. COLEC11 binds to various sugars and LPS (lipopolysaccharides), which include fucose but does not bind to glucose, hnRNP, Beta-1,3-Gal-T3 or mannose. COLEC11 is ubiquitously expressed in most tissues with high expression in kidney, liver, fetal liver, small intestine, thymus, spinal cord, placenta, adrenal gland, pancreas and several cell lines. COLEC11 is a secreted protein and all alternatively spliced isoforms of COLEC11 have oligomeric structures created through disulfide bonding.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COLEC11
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge