Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PLEKHM1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8062R-A680
BOSSBS-8062R-A680EA 893 EUR InStock
BOSSBS-8062R-A680
Anti-PLEKHM1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Involved in vesicular transport in the osteoclast (By similarity). May have a role in sialyl-lex-mediated transduction of apoptotic signals.Tissue specificity: Expressed in placenta, liver, prostate, thymus, spleen, ovary, colon, colon carcinoma and peripheral blood lymphocytes (PBL). Weakly expressed in brain, lung, kidney, and testis. No expression in heart, skeletal muscle, pancreas and small intestine. Predominantly expressed in the breast carcinoma cell line MCF-7.Involvement in disease:Defects in PLEKHM1 are the cause of osteopetrosis autosomal recessive type 6 (OPTB6); also known as autosomal recessive osteopetrosis intermediate form. Osteopetrosis is a rare genetic disease characterised by abnormally dense bone, due to defective resorption of immature bone. The disorder occurs in two forms: a severe autosomal recessive form occurring in utero, infancy, or childhood, and a benign autosomal dominant form occurring in adolescence or adulthood. Autosomal recessive osteopetrosis is usually associated with normal or elevated amount of non-functional osteoclasts.

Type: Primary
Antigen: PLEKHM1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 546-700/1056
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge