Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PLEKHM3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8064R-A647
BOSSBS-8064R-A647EA 893 EUR InStock
BOSSBS-8064R-A647
Anti-PLEKHM3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
PLEKHM3 (pleckstrin homology domain containing, family M, member 3), also known as DAPR or PLEKHM1L (pleckstrin homology domain containing, family M, member 1-like), is a 761 amino acid phosphoprotein that contains two pleckstrin homology (PH) domains and one phorbol-ester/DAG-type zinc finger. Conserved in chimpanzee, dog, cow, mouse, rat, chicken and zebrafish, PLEKHM3 exists as three alternatively spliced isoforms that participate in metal ion binding. The gene that encodes PLEKHM3 maps to human chromosome 2q33.3. As the second largest human chromosome, chromosome 2 makes up approximately 8% of the human genome and contains 237 million bases encoding over 1,400 genes. Chromosome 2 contains a probable vestigial second centromere, as well as vestigial telomeres, which gives credence to the hypothesis that human chromosome 2 formed as a result of an ancient fusion of two ancestral chromosomes, which are still present in modern day apes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLEKHM3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge