Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CCDC34 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8098R-A350
BOSSBS-8098R-A350EA 893 EUR InStock
BOSSBS-8098R-A350
Anti-CCDC34 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The coiled-coil domain is a structural motif found in proteins that are involved in a diverse array of biological functions such as the regulation of gene expression, cell division, membrane fusion and drug extrusion and delivery. CCDC34 (coiled-coil domain containing 34), also known as Renal carcinoma antigen NY-REN-41, is a 373 amino acid protein that is expressed in testis, breast, lung, placenta, liver and small intestine. A translocation between the short arms of chromosomes 11 and 18 affecting the CCDC34 gene has been identified in a patient with hamartoma of the retinal pigment epithelium. There are two isoforms of CCDC34 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC34
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge