Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CCDC117 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8112R-A647
BOSSBS-8112R-A647EA 893 EUR InStock
BOSSBS-8112R-A647
Anti-CCDC117 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
CCDC117 is a 279 amino acid protein that is expressed as multiple alternatively spliced isoforms and is encoded by a gene which maps to human chromosome 22. Chromosome 22 houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, Neurofibromatosis type 2, autism and schizophrenia. Additionally, translocations between chromosomes 9 and 22 may lead to the formation of the Philadelphia chromosome and the subsequent production of the novel fusion protein Bcr-Abl, a potent cell proliferation activator found in several types of leukemias.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC117
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge