Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CCDC88B Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8141R-CY5.5
BOSSBS-8141R-CY5.5EA 900 EUR InStock
BOSSBS-8141R-CY5.5
Anti-CCDC88B Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
HkRP3, also known as CCDC88B (coiled-coil domain-containing protein 88B) or BRLZ (brain leucine zipper domain-containing protein), is a 1,476 amino acid protein that belongs to the CCDC88 family. Members of the hook-related protein family are characterized by the presence of a C-terminal hook-related domain and an N-terminal potential microtubule binding domain. HkRP3 may be involved in the linkage of various organelles to microtubules, and exists as six alternatively spliced isoforms. The gene encoding HkRP3 maps to human chromosome 11q13.1 and mouse chromosome 19 A. Chromosome 11 houses over 1,400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC88B
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge