Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FRMD5 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8237R-CY3
BOSSBS-8237R-CY3EA 900 EUR InStock
BOSSBS-8237R-CY3
Anti-FRMD5 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
FERM domains are roughly 150 amino acids in length and are found in a number of cytoskeletal-associated proteins such as Ezrin, Radixin, Moesin and 4.1 (erythrocyte membrane protein band 4.1), where they provide a link between cytoskeletal signals and membrane dynamics. FRMD5 (FERM domain-containing protein 5) is a 570 amino acid single-pass membrane protein that contains one FERM domain and exists as two alternatively spliced isoforms. The gene encoding FRMD5 maps to human chromosome 15, which houses over 700 genes and comprises nearly 3% of the human genome. Angelman syndrome, Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are all associated with defects in chromosome 15-localized genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FRMD5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge