Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CA11 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8244R-A350
BOSSBS-8244R-A350EA 893 EUR InStock
BOSSBS-8244R-A350
Anti-CA11 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes responsible for catalyzing the reversible hydration of carbon dioxide. CAs show extensive diversity in their distribution and subcellular localization. They are involved in a variety of biological processes, including calcification, bone resorption, respiration, acid-base balance and the formation of aqueous humor, saliva, gastric juice and cerebrospinal fluid. CA XI, also referred to as carbonic anhydrase-related protein 11 precursor (CA-RP XI) or carbonic anhydrase-related protein 2 (CA-RP II), is a member of the carbonic anhydrase family that lacks two of the three Zn-binding motifs essential for carbonic anhydrase activity. For this reason, CA XI does not exhibit catalytic activity. It is expressed primarily in brain but is also found in spinal cord and thyroid. CA XI may play a role in brain development.Tissue specificity:Expressed abundantly in the brain with moderate expression also present in spinal cord and thyroid.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CA11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge