Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DHRS13 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8261R
BOSSBS-8261REA 725 EUR InStock
BOSSBS-8261R
Anti-DHRS13 Rabbit Polyclonal Antibody
Antikörper
DHRS13 (dehydrogenase/reductase SDR family member 13), also known as UNQ419/PRO853, is a 377 amino acid secreted protein belonging to the short-chain dehydrogenases/reductases (SDR) family. DHRS13 is presumed to function as an oxidoreductase and is phosphorylated, potentially by ATM or ATR, upon DNA damage. DHRS13 exists as two isoforms produced by alternative splicing events. The gene encoding DHRS13 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes. Neurofibromatosis, dysregulated Schwann cell growth, Alexander disease, Birt-Hogg-Dube syndrome and Canavan disease are associated with chromosome 17.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DHRS13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge