Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GIMAP3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8269R-CY5.5
BOSSBS-8269R-CY5.5EA 900 EUR InStock
BOSSBS-8269R-CY5.5
Anti-GIMAP3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
GIMAP3p is a murine mitochondrial membrane protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. The IAN/GIMAP family consists of GTP-binding proteins that share a unique primary structure and whose expression is finely regulated by T-cell receptor signals. Recent studies have shown that IAN/GIMAP family proteins crucially regulate the survival of T cells during development, selection and homeostasis, and are possibly linked to the onset of T-lymphopenia, leukemia and autoimmunity. IAN/GIMAP family proteins might also take part in mitochondrial regulation of lymphocyte apoptosis by interacting with Bcl-2 family proteins. The human GIMAP (GTPase of the immunity-associated protein) gene family includes seven functional members residing on (human) chromosome 7q36.1 and one pseudogene (hGIMAP3).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GIMAP3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge