Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GIMAP5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8271R-HRP
BOSSBS-8271R-HRPEA 900 EUR InStock
BOSSBS-8271R-HRP
Anti-GIMAP5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
GIMAP5, also known as IAN5, IAN4L1 (immunity-associated nucleotide 4-like 1 protein), IAN4, IMAP3, HIMAP3 or IROD, is a 307 amino acid single-pass type IV membrane protein of the mitochondrial outer membrane. A member of the GTP-binding superfamily and the immuno-associated nucleotide (IAN) subfamily, GIMAP5 plays a role in T-cell survival and mitochondrial integrity. GIMAP5 is highly expressed in CD4 and CD8-positive T-cells and monocytes, as well as B-lymphocyte-derived neoplasms. GIMAP5 exists as two alternatively spliced isoforms and is encoded by a gene that maps to human chromosome 7q36.1. Defects in the GIMAP5 gene are associated with systemic lupus erythematosus.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: GIMAP5
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge