Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EP400 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8279R
BOSSBS-8279REA 725 EUR InStock
BOSSBS-8279R
Anti-EP400 Rabbit Polyclonal Antibody
Antikörper
NuAR is a multi-protein histone acetyltransferase complex that functions to acetylate the nucleosomal Histones H4 and H2A, thereby activating transcription of select target genes. p400, also known as EP400 (E1A binding protein p400) or mDomino, localizes to the nucleus and is a component of the NuAR complex. Expressed in brain, liver, thymus, lung, spleen, colon and kidney, p400 regulates the transcriptional activity of proteins such as MZF-1 and contributes to the ATPase and helicase activities of NuA4. p400 is a SWI2/ SNF2-related protein that can interact with the adenovirus oncoprotein E1A, thus activating the proapoptotic activity of E1A. The ability of p400 to regulate transcriptional and apoptotic activity suggests that the NuAR complex may be a crucial component of cell proliferation, transformation and, possibly, carcinogenesis. Five isoforms of p400 exist due to alternative splicing events.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: EP400
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge