Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DPH2 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8282R
BOSSBS-8282REA 725 EUR InStock
BOSSBS-8282R
Anti-DPH2 Rabbit Polyclonal Antibody
Antikörper
DPH2 (diphthamide biosynthesis protein 2), also known as DPH2L2, is a 489 amino acid protein that shows strong expression in skeletal muscle; moderate expression in heart, small intestine, liver, pancreas, testis and colon; and lesser expression in brain, placenta, kidney, spleen, thymus, prostate, ovary and lymphocytes. DPH2 interacts with DPH1 and, functioning together as a dimer or multimer, DPH1 and DPH2 may participate in diphthamide biosynthesis. Diphthamide is a posttranslationally modified histidine residue which occurs in EF-2 (elongation factor 2) and targets diphtheria toxin ADP-ribosylation. The loss of DPH2 in Saccharomyces cerevisiae is believed to suppress zymocicity. Two transcript variants encoding different isoforms have been found for this gene.Required for the first step in the synthesis of diphthamide, a post-translational modification of histidine which occurs in translation elongation factor 2.Tissue specificity:Strongly expressed in skeletal muscle. Moderately expressed in heart, small intestine, liver, pancreas, testis and colon. Weakly expressed in brain, placenta, kidney, speen, thymus, prostate, ovary and lymphocytes.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DPH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge