Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DYDC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8286R-A647
BOSSBS-8286R-A647EA 893 EUR InStock
BOSSBS-8286R-A647
Anti-DYDC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
The Dumpy-30 (DPY-30) protein was first described in C. elegans, in which it is involved in dosage compensation of sex chromosomes. Conserved from yeast to humans, the DPY-30 family is involved in gene expression and chromatin modification, specifically histone methylation. DPY-30 and closely related proteins contain a short motif that is related to the dimerization motif in the regulatory subunit of protein kinase A (PKA), which consists of two ?helices that form a four-helix bundle during dimerization. As a member of the DPY-30 family, DYDC1 (DPY30 domain-containing protein 1), also known as DPY30D1 and RSD9, is a 177 amino acid protein that binds to Endophilin III and plays a crucial role during acrosome biogenesis. DYDC1 is specifically expressed in brain and testis and accumulates in the acrosome area during spermatogenesis. Knockdown of DYDC1 mRNA results in disruption of acrosome formation and spermatid differentiation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DYDC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge