Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PRDX2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8300R-A488
BOSSBS-8300R-A488EA 893 EUR InStock
BOSSBS-8300R-A488
Anti-PRDX2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Peroxiredoxin (Prx) is an antioxidant enzyme detoxifying reactive oxygen species and has a cysteine at the active site. Prx enzymes modulate various receptor signaling pathways and protect cells from oxidatively induced death. Peroxiredoxin 1 to 4 have two conserved Cys residues corresponding to Cys51 and Cys172 of mammalian Peroxiredoxin 1. The active site cysteine(Cys51) is oxidized to cysteine sulfenic acid(Cys51-SOH) when a peroxide is reduced. Because Cys51-SOH is unstable, it forms a disulfide with Cys172-SH which comes from the other subunit of the homodimer. The disulfide is then reduced back to the Prx active thiol form by the thioredoxin-thioredoxin reductase system. However, the formation of the disulfide is a slow process. Thus under oxidative stress conditions, the sulfenic intermediate(Cys51-SOH) can be easily over oxidized to cysteine sulfinic acid(Cys-SO2H) or cysteine sulfonic acid(Cys-SO3H) before it is able to form a disulfide. Recent studies suggest that over oxidized Prx can be reduced back to the active form during recovery after oxidative stress.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRDX2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge