Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GLT8D2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8302R-A488
BOSSBS-8302R-A488EA 893 EUR InStock
BOSSBS-8302R-A488
Anti-GLT8D2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
GLT8D2 (glycosyltransferase 8 domain-containing protein 2), also known as GALA4A, is a 349 amino acid single-pass type II membrane protein. A member of the glycosyltransferase 8 family, GLT8D2 is encoded by a gene that maps to human chromosome 12q23.3. Encoding over 1,100 genes within 132 million base pairs, chromosome 12 makes up about 4.5% of the human genome. A number of skeletal deformities are linked to chromosome 12, including hypochondrogenesis, achondrogenesis, Noonan syndrome, Kniest dysplasia and trisomy 12p. Chromosome 12 is also home to a homeobox gene cluster, which encodes crucial transcription factors for morphogenesis, as well as the natural killer complex gene cluster, which encodes C-type lectin proteins that mediate the NK cell response to MHC I interaction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GLT8D2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge