Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HDHD3 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8318R-CY7
BOSSBS-8318R-CY7EA 900 EUR InStock
BOSSBS-8318R-CY7
Anti-HDHD3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
HDHD3 (haloacid dehalogenase-like hydrolase domain containing 3) is a 251 amino acid protein that belongs to the HAD-like hydrolase superfamily, which contains a group of hydrolase enzymes that differ from the å/∫ hydrolase family based on structure. This family of hydrolase enzymes includes L-2-haloacid dehalogenase, epoxide hydrolases and phosphatases. HDHD3 is encoded by a gene located on human chromosome 9q32. Chromosome 9 consists of about 145 million bases, 4% of the human genome and encodes nearly 900 genes. Thought to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HDHD3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge