Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PA26 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8325R-A555
BOSSBS-8325R-A555EA 893 EUR InStock
BOSSBS-8325R-A555
Anti-PA26 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Cell cycle progression is subject to arrest at G1 and G2 checkpoints in response to DNA damage, presumably to allow time for DNA repair prior to entry into S and M phase, respectively. The p53 tumor suppressor is required for one such G1 checkpoint and functions to upregulate expression of GADD 45 and the mitotic inhibitory protein p21. GADD 45 stimulates DNA excision repair in vitro and inhibits entry of cells into S phase, and it apparently acts in concert with GADD 153 in inducing growth arrest. A related DNA-damage inducible gene, GADD 34 synergizes with GADD 45 or GADD 153 in suppressing cell growth. PEG-3 (progression elevated gene-3) shares significant homology with GADD 34 and is inducible by DNA damage. An additional GADD related gene, PA26, is a possible target of p53. Three isoforms of PA26 have been identified as PA26-T1, PA26-T2 and PA26-T3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PA26
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge