Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PANK3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8339R-FITC
BOSSBS-8339R-FITCEA 900 EUR InStock
BOSSBS-8339R-FITC
Anti-PANK3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
The pantothenate kinase (PANK) family of proteins catalyzes the first step in coenzyme A (CoA) biosynthesis. Coenzyme A is an important coenzyme involved in the synthesis and oxidation of fatty acids, as well as the oxidation of pyruvate in the citric acid (Krebs) cycle. Pantothenate kinase 3 (PANK3) is a 370 amino acid member of the pantothenate kinase family that plays a role in the physiological regulation of the intracellular CoA concentration. Localized to the cytoplasm, PANK3 is regulated by feedback inhibition by CoA and its thioesters. PANK3 transfers a phosphate from ATP to pantothenate (Vitamin B5), resulting in formation of 4’-phosphopantothenate. Closely related to its family members, PANK1, PANK2 and PANK4, PANK3 is highly expressed in liver. Pantothenate kinase associated neurodegeneration (PKAN) results from mutations in the gene encoding PANK2, the only mitochondria targeted human PANK.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PANK3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge