Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CPXM1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8341R-A350
BOSSBS-8341R-A350EA 893 EUR InStock
BOSSBS-8341R-A350
Anti-CPXM1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
CPXM (carboxypeptidase X, member 1) belongs to the peptidase M14 family. However, no carboxypeptidase activity has yet been detected. It may be involved in cell-cell interactions.Members of the M14 metallocarboxypeptidase protein family serve many diverse functions and are divided into three subfamilies based on structure, function and amino acid sequence similarity. Belonging to the N/E subfamily, CPXM (metallocarboxypeptidase CPX-1) is a 734 amino acid protein that contains a F5/8 type C domain and likely binds one zinc ion per subunit. Most members of the N/E subfamily contain several domains, including an active carboxypeptidase domain and signal peptide, and are thought to function mostly in protein-protein interactions and/or protein-membrane interactions, thereby targeting the protein to specific locations within the secretory pathway. CPXM is a unique member of this subfamily in that it does not appear to exhibit any enzymatic activity due to lack of several active-site residues that are present in the catalytic domain of other members of the N/E subfamily. Studies showing that CPXM expression is regulated during osteoclastogenesis suggest that CPXM may play a role in osteoclast differentiation. There are two isoforms of CPXM which are a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CPXM1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge