Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FGFR1OP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8345R-FITC
BOSSBS-8345R-FITCEA 900 EUR InStock
BOSSBS-8345R-FITC
Anti-FGFR1OP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
FOP, is a 399 amino acid protein that localizes to the centrosome and contains one LisH domain. Expressed ubiquitously with highest expression in kidney, heart, muscle, colon, liver, testis and pancreas, FOP functions as a homodimer that interacts with EB1 and CEP350 and is essential for anchoring microtubules to centrosomes. Chromosomal aberrations that involve the FOP gene are associated with the pathogenesis of stem cell myeloproliferative disorder (MPD), a condition that is characterized by eosinophilia and myeloid hyperplasia and ultimately leads to acute myeloid leukemia. FOP is expressed as multiple isoforms that are produced by alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGFR1OP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge