Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FGFR1OP2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8346R-CY5.5
BOSSBS-8346R-CY5.5EA 900 EUR InStock
BOSSBS-8346R-CY5.5
Anti-FGFR1OP2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
FGFR1OP2 belongs to the SIKE family. The FGFR1OP2 (FGFR1 oncogene partner 2) gene was identified through its involvement in a fusion with the FGFR1 gene. FGFR1OP2 may be involved in the wound healing pathway. It is expressed in bone marrow, spleen and thymus. A chromosomal aberration involving FGFR1OP2 may be a cause of stem cell myeloproliferative disorder (MPD). Insertion ins(12;8)(p11;p11p22) with FGFR1. MPD is characterized by myeloid hyperplasia, eosinophilia and T cell or B cell lymphoblastic lymphoma. In general it progresses to acute myeloid leukemia. The fusion protein FGFR1OP2-FGFR1 may exhibit constitutive kinase activity and be responsible for the transforming activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGFR1OP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge