Anti-NIPA Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-8363R-CY5.5
BOSSBS-8363R-CY5.5EA 900 EUR InStock
BOSSBS-8363R-CY5.5
Anti-NIPA Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The regulated oscillation of protein expression is an essential mechanism of cell cycle control. The SCF class of E3 ubiquitin ligases is involved in this process by targeting cell cycle regulatory proteins for degradation by the proteasome, with the F-box subunit of the SCF specifically recruiting a given substrate to the SCF core. NIPA (nuclear interaction partner of ALK) is a human F-box-containing protein that defines an SCF-type E3 ligase (SCFNIPA) controlling mitotic entry. Assembly of this SCF complex is regulated by cell-cycle-dependent phosphorylation of NIPA, which restricts substrate ubiquitination activity to interphase. Nuclear cyclin B1 is a substrate of SCFNIPA. Inactivation of NIPA by RNAi results in nuclear accumulation of cyclin B1 in interphase, activation of cyclin B1-Cdk1 kinase activity, and premature mitotic entry. Thus, SCFNIPA-based ubiquitination may regulate S-phase completion and mitotic entry in the mammalian cell cycle.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NIPA
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge