Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-UBE2Q1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8383R-A647
BOSSBS-8383R-A647EA 893 EUR InStock
BOSSBS-8383R-A647
Anti-UBE2Q1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (UBE1s), ubiquitin-conjugating enzymes (UBE2s), and ubiquitin-protein ligases (UBE3s). When ubiquitin is activated by a UBE1, it is transferred to the cysteine residue on a UBE2. UBE2 then binds a UBE3, which transfers the ubiquitin from the UBE2 cysteine to a lysine residue on the target protein. Ubiquitin-conjugating enzyme E2 Q1 (UBE2Q1), also known as ubiquitin-protein ligase Q1 or ubiquitin carrier protein Q1, is an 422 amino acid protein belonging to the ubiquitin-conjugating enzyme (UBE2) family. Two named isoforms of UBE2Q1 exist as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UBE2Q1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge