Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CBLL1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8386R-A647
BOSSBS-8386R-A647EA 890 EUR InStock
BOSSBS-8386R-A647
Anti-CBLL1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
CBLL1, also known as HAKAI (meaning ‘destruction’ in Japanese), or RNF188 (RING finger protein 188), is a 491 amino acid protein that contains one C2H2-type zinc finger and one RING-type zinc finger. CBLL1 is believed to function as an E3 ubiquitin-protein ligase that accepts a ubiquitin residue from an E2 ubiquitin-conjugating enzyme and immediately transfers that residue to a protein that is targeted for degradation. More specifically, upon activation of c-Src, CBLL1 interacts with and ubiquitinates tyrosine-phosphorylated E-cadherin, thereby targeting the E-cadherin complex for endocytosis and disrupting epithelial cell-cell contacts. Via its role as an E-cadherin regulator, CBLL1 participates in cell adhesion and may also be involved in the regulation of epithelial-mesenchymal transitions.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CBLL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge