Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FBXW2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8393R-CY3
BOSSBS-8393R-CY3EA 900 EUR InStock
BOSSBS-8393R-CY3
Anti-FBXW2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
F box proteins are an expanding family of eukaryotic proteins characterized by an approximately 40 amino acid motif, the F box. Some F box proteins have been shown to be critical for the ubiquitin mediated degradation of cellular regulatory proteins. In fact, F box proteins are one of the four subunits of ubiquitin protein ligases, called SCFs. SCF ligases bring ubiquitin conjugating enzymes to substrates that are specifically recruited by the different F box proteins. A large family of mammalian F box proteins has recently been identified and classified into three groups based on the presence of either the WD 40 repeats, the leucine rich repeats, or the presence or absence of other protein protein interacting domains. The FBXW2 gene product, the second identified member of the F box gene family, contains multiple WD 40 repeats.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FbxW2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge