Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FBXW8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8395R-A680
BOSSBS-8395R-A680EA 893 EUR InStock
BOSSBS-8395R-A680
Anti-FBXW8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
FBXO29, also designated F-box/WD repeat-containing protein 8 (FBXW8), is a 598 amino acid protein that contains one 40 amino acid F-box region, making it a member of the F-box family. FBXO29 also contains five WD repeats. F-box proteins are critical components of the SCF (Skp1-CUL-1-F-box protein) type E3 ubiquitin ligase complex and are involved in substrate recognition and recruitment for ubiquitination. F-box proteins are members of a large family that regulates cell cycle, immune response, signaling cascades and developmental programs by targeting proteins, such as cyclins, cyclin-dependent kinase inhibitors, IkB-å and -catenin, for degradation by the proteasome after ubiquitination. Functioning as a component of the SCF complex, FBXO29 is thought to recognise and bind to select phosphorylated proteins, thereby promoting their ubiquitination and subsequent degradation. FBXO29 exists as two isoforms as a result of alternative splicing events.

Type: Primary
Antigen: FBXW8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 181-280/598
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge