Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HEXDC Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8418R-CY5
BOSSBS-8418R-CY5EA 900 EUR InStock
BOSSBS-8418R-CY5
Anti-HEXDC Rabbit Polyclonal Antibody (Cy5®)
Antikörper
HEXDC, also known as hexosaminidase D, beta-hexosaminidase D, N-acetyl-beta-galactosaminidase, hexosaminidase domain-containing protein or beta-N-acetylhexosaminidase, is a 486 amino acid cytoplasmic and nuclear protein that has hexosaminidase activity and belongs to the glycosyl hydrolase 20 family. Existing as two alternatively spliced isoforms, HEXDC catalyzes the hydrolysis of non-reducing N-acetyl-D-hexosamine residues near the termini of N-acetyl-beta-D-hexosaminides. The gene encoding HEXDC maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Defects in p53 is associated with malignant cell growth and Li-Fraumeni syndrome. BRCA1 is directly involved in DNA repair and is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HEXDC
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge