Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZNF537 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8540R-CY5
BOSSBS-8540R-CY5EA 900 EUR InStock
BOSSBS-8540R-CY5
Anti-ZNF537 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The homeobox DNA-binding domain is a 60 amino acid motif that is conserved among many species and functions to bind DNA via a helix-turn-helix structure, thereby playing a role in transcriptional regulation and the control of gene expression. TSHZ3 (teashirt zinc finger homeobox 3), also known as KIAA1474, TSH3 or ZNF537, is a 1,081 amino acid protein that contains one homeobox DNA-binding domain and five C2H2-type zinc fingers. Localized to the nucleus, TSHZ3 functions as a transcriptional regulator that is involved in developmental processes throughout the body. The gene encoding TSHZ3 maps to human chromosome 19, which consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF537
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge