Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IGF2BP2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8559R-CY5
BOSSBS-8559R-CY5EA 1350 EUR InStock
BOSSBS-8559R-CY5
Anti-IGF2BP2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
IGF2BP2 (insulin-like growth factor 2 mRNA binding protein 2) is also known as IGF2 mRNA-binding protein 2, IMP-2 (IGF-II mRNA-binding protein 2), VICKZ family member 2 or hepatocellular carcinoma autoantigen p62 and is a 556 amino acid protein. IGF2BP2 is expressed in a variety of tissues including heart, placenta, skeletal muscle, pancreas, fetal liver, lung, kidney, thymus and gonadal cells. IGF2BP2 is an RNA binding protein which may be involved in the regulation of mRNA translation and may also function to control the spatial localization of target mRNAs. against IGF2BP2 have been detected in patients with HCC (hepatocellular carcinoma), suggesting that IGF2BP2 may have a role in the pathogenesis of HCC. Defects in IGF2BP2 are thought to be associated with susceptibility to type 2 diabetes mellitus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IGF2BP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge