Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MLLT11 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8562R-CY5.5
BOSSBS-8562R-CY5.5EA 900 EUR InStock
BOSSBS-8562R-CY5.5
Anti-MLLT11 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The gene encoding the Mixed-Lineage Leukemia (MLL) proteins is located on chromosome 11q23. Chromosomal translocations involving band 11q23 result in rogue activator proteins that are associated with approximately 10% of patients with acute lymphoblastic leukemia (ALL) and 5% of patients with acute myeloid leukemia (AML). Most patients affected are less than 1 year of age. MLLT11, also known as mixed-lineage leukemia translocated to 11 or AF1q, is a 90 amino acid MLL fusion partner. Based on the expression patterns of MLLT11, it is thought that MLLT11 plays a role in leukemogenesis and, specifically, the progression of acute monocytic leukemia (AML). Also, expressed in embryonic brain cortex, MLLT11 is upregulated during neuronal differentiation and is thought to play a role in the development of the central nervous system. Finally, MLLT11 has been shown to be differentially expressed in highly metastatic cells, in comparison with non-metastatic parent cells. Such findings suggest a role of MLLT11 in tumorigenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MLLT11
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge