Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RENT1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8565R-CY3
BOSSBS-8565R-CY3EA 900 EUR InStock
BOSSBS-8565R-CY3
Anti-RENT1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
In eukaryotes, it is essential to have the ability to detect and degrade transcripts that lack full coding potential. Nonsense-mediated RNA decay (NMD) protects the organism by avoiding the translation of truncated peptides with dominant negative or deleterious gain-of-function potential. Rent1, a mammalian ortholog of Upflp, is essential for embryonic viability (1–3). Rent1 (also designated regulator of nonsense transcripts and HUpf1) contains an N-terminal zinc finger-like domain, NTPase domains and a region comprised of domains that define Rent1 as a superfamily group I helicase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RENT1/hUPF1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge