Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GPKOW Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8577R-A350
BOSSBS-8577R-A350EA 893 EUR InStock
BOSSBS-8577R-A350
Anti-GPKOW Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
GPKOW is a potential RNA-binding protein consisting of one central G patch domain and two C-terminal KOW domains. T54 is a 476 amino acid protein belonging to the MOS2 family. It is a mammalian homolog of the Arabidopsis thaliana MOS2 (modifier of SNC1, 2) nuclear protein that is required for innate immunity. Similar to A. thaliana MOS2, T54 localizes to the nucleus and contains G patch and KOW domains, suggesting that T54 may play a similar role in mammalian innate immunity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPKOW
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge