Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GTDC1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8582R-A680
BOSSBS-8582R-A680EA 893 EUR InStock
BOSSBS-8582R-A680
Anti-GTDC1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Glycosyltransferases that mediate the regio- and stereoselective transfer of sugars, such as the fucosyltransferases, determine cell surface-carbohydrate profiles, which is an essential interface for biological recognition processes. GTDC1 (Glycosyltransferase-like domain-containing protein 1), also known as Mat-Xa, is a 458 amino acid protein belonging to the glycosyltransferase 1 family. GTDC1 is ubiquitously expressed, with highest levels found in peripheral blood leukocytes, spleen, lung and testis.An extremely rare recessive genetic disorder, Alstr syndrome, is caused by mutations in the ALMS1 gene, which maps to chromosome 2.

Type: Primary
Antigen: GTDC1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge