Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ETV5 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8592R-CY3
BOSSBS-8592R-CY3EA 922 EUR InStock
BOSSBS-8592R-CY3
Anti-ETV5 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Rad and Gem related GTP binding protein (REM) is a member of the RGK subfamily of Ras-like GTPases that also includes Rad, REM2 and Gem/Kir. REM is a phosphorylated protein that is highly expressed in cardiac muscle and moderately expressed in lung, kidney and skeletal muscle. REM associates with several 14-3-3 isoforms as well as with calmodulin in a calcium-dependent manner. REM mediates two distinct signal transduction pathways that regulate both cytoskeletal reorganization and voltage-gated calcium channel activity. REM decreases the current that passes through cardiac voltage-gated L-type Ca channels (Ca(V)). Overexpression of REM may result in the development of cytoplasmic processes, reorganization of the Actin cytoskeleton, reduction in focal adhesion size and an elongated or dendritic-like cell morphology.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ETV5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge