Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GPR120 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8596R-CY5
BOSSBS-8596R-CY5EA 897 EUR InStock
BOSSBS-8596R-CY5
Anti-GPR120 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
GPR120, a member of the rhodopsin family of G protein-coupled receptors (GPCRs), is a 377 amino acid protein which is expressed in the intestine. GPR120 is a receptor for unsaturated long-chain FFAs (free fatty acids). FFAs act as signaling molecules and are an important energy source. They also employ various physiological responses through their GPCRs. One such response occurs when dietary FFAs stimulate GPR120. This stimulation promotes the secretion of glucagon-like peptide 1 (GLP-1) in vivo and in vitro. GLP-1 belongs to the class of molecules known as the incretins, which are associated with insulin secreted from the pancreas as a result of food intake. GLP-1 also inhibits glucagon and gastric acid secretion and gastric emptying. Consequently, the role of GPR120 in the secretion of GLP-1 is critical in the treatment of diabetes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPR120
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge