Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SLC9A3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8601R-CY5
BOSSBS-8601R-CY5EA 900 EUR InStock
BOSSBS-8601R-CY5
Anti-SLC9A3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
NHE-3 are integral membrane proteins that are expressed in most mammalian tissues, where they regulate intracellular pH and cell volume. NHEs mediate the transport of hydrogen (H+) ions out of cells in exchange for extracellular sodium (Na+) ions. While NHE-1 is ubiquitously expressed, the NHE isoforms 2-8 have distinct tissue- and cell type-dependent expression and inhibitory characteristics. NHE-3 localizes to the apical membrane of renal proximal tubules where it is responsible for most of the sodium transport and fluid reabsorption. NHE-3 translocates to internal pools where it mediates natriuresis when blood pressure increases abruptly. NHE-3 is also expressed in the stomach and functions to protect the mucosa by secreting protons that diffuse into the mucous cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC9A3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge