Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BASP1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8662R-A555
BOSSBS-8662R-A555EA 893 EUR InStock
BOSSBS-8662R-A555
Anti-BASP1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Neuronal axonal membrane protein Nap-22, also designated neuronal tissue-enriched acidic protein or brain acid soluble protein (BASP1), is a Ca2+-dependent calmodulin-binding protein that is important for neuronal sprouting and plasticity. Nap-22 is abundant in brain nerve terminals and is also present in significant amounts in kidney, testis and lymphoid tissue. Nap-22 undergoes N-terminal myristoylation for membrane localization. It has been characterized as a major protein of neuronal rafts, which are known to preferentially bind membranes containing cholesterol. Nap-22 is a crucial protein active in neurite outgrowth and synaptic plasticity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BASP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge