Anti-MEPE Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-8689R-A750
BOSSBS-8689R-A750EA 893 EUR InStock
BOSSBS-8689R-A750
Anti-MEPE Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
MEPE is a 525 amino acid extracellular matrix protein. Expressed in osteocytes and brain, MEPE is a regulator of bone metabolism that is thought to mediate mineralisation and demineralisation within the osteocyte microenvironment. MEPE contains an RGD cell-attachment motif and shares molecular similarities with several dentin-bone extracellular matrix RGD-containing phosphoglycoproteins, including OPN (osteopontin) and DSP (dentin sialophosphoprotein). Via its ability to control bone mineralisation, MEPE is associated with various developmental events such as skeletogenesis, bone regeneration and odontogenesis. MEPE is secreted in hypophosphatemic osteomalacia tumors, suggesting a possible role in the pathophysiology of bone-related cancers.
Type: Primary
Antigen: MEPE
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/525
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge